missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic Dihydrazide 90.0+%, TCI America™

Catalog No. T075825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T075825G 25 g
T0758500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T075825G Supplier TCI America Supplier No. T075825G

Chemical Identifiers

CAS 136-64-1
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00014762
InChI Key ALHNLFMSAXZKRC-UHFFFAOYSA-N
Synonym terephthalic dihydrazide, terephthalohydrazide, terephthalic acid dihydrazide, terephthalic hydrazide, 1,4-benzenedicarboxylic acid, dihydrazide, terephthaloyl dihydrazide, terephthalic acid bishydrazide, terephthalic acid, dihydrazide, terephthalhydrazide, terephthalicdihydrazide
PubChem CID 67294
IUPAC Name benzene-1,4-dicarbohydrazide
SMILES NNC(=O)C1=CC=C(C=C1)C(=O)NN

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalic Dihydrazide