accessibility menu, dialog, popup

UserName

CAS RN 136-64-1

IUPAC Name: benzene-1,4-dicarbohydrazide
Synonyms: benzene-1,4-dicarbohydrazide
Molecular Weight (g/mol): 194.19
Molecular Formula: C8H10N4O2
InChi Key: ALHNLFMSAXZKRC-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(C=C1)C(=O)NN
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)