missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic dihydrazide, 95%

Catalog No. AAL0101706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0101706 5 g
AAL0101714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0101706 Supplier Thermo Scientific Chemicals Supplier No. L0101706

CAS: 136-64-1 | C8H10N4O2 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136-64-1
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00014762
InChI Key ALHNLFMSAXZKRC-UHFFFAOYSA-N
Synonym terephthalic dihydrazide, terephthalohydrazide, terephthalic acid dihydrazide, terephthalic hydrazide, 1,4-benzenedicarboxylic acid, dihydrazide, terephthaloyl dihydrazide, terephthalic acid bishydrazide, terephthalic acid, dihydrazide, terephthalhydrazide, terephthalicdihydrazide
PubChem CID 67294
IUPAC Name benzene-1,4-dicarbohydrazide
SMILES NNC(=O)C1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 982939
Formula Weight 194.19
Percent Purity 95%
Chemical Name or Material Terephthalic dihydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-253-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only