missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic Acid 99.0+%, TCI America™

Catalog No. p-7136115
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
T016625G 25 g
T0166500G 500 g
2 options

Catalog No. T016625G

Supplier: TCI America T016625G

Only null left

Chemical Identifiers

CAS 100-21-0
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002558
InChI Key KKEYFWRCBNTPAC-UHFFFAOYSA-N
Synonym p-phthalic acid, 1,4-benzenedicarboxylic acid, benzene-1,4-dicarboxylic acid, p-dicarboxybenzene, p-benzenedicarboxylic acid, p-carboxybenzoic acid, acide terephtalique, para-phthalic acid, tephthol, 1,4-dicarboxybenzene
PubChem CID 7489
ChEBI CHEBI:15702
IUPAC Name terephthalic acid
SMILES C1=CC(=CC=C1C(=O)O)C(=O)O

Specifications

Melting Point 402°C
Color White
Quantity 25 g
Formula Weight 166.13
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalic Acid
EINECSNumber (3)-1334
RTECSNumber WZ0875000
TSCA Yes