missing translation for 'onlineSavingsMsg'
Learn More

Terephthaldicarboxaldehyde, 98%

CAS: 623-27-8 | C8H6O2 | 134.13 g/mol

$482.48 - $482.48

Chemical Identifiers

CAS 623-27-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00006949
InChI Key KUCOHFSKRZZVRO-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde
PubChem CID 12173
IUPAC Name terephthalaldehyde
SMILES C1=CC(=CC=C1C=O)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC137895000
SDS View Documents
Thermo Scientific Chemicals
137895000
500 g Plastic bottle
Each for $482.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

623-27-8
134.13
KUCOHFSKRZZVRO-UHFFFAOYSA-N
12173
C1=CC(=CC=C1C=O)C=O
C8H6O2
MFCD00006949
1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde
terephthalaldehyde

Specifications

623-27-8
White to Yellow
76°C
98%
C8H6O2
MFCD00006949
07, 675
Solubility in water: 3g/L (50°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
C1=CC(=CC=C1C=O)C=O
134.13
134.13
Crystalline Powder
114°C to 116°C
245°C to 248°C
Authentic
Plastic bottle
C6H4(CHO)2
500 g
1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde
KUCOHFSKRZZVRO-UHFFFAOYSA-N
terephthalaldehyde
12173
98%
Terephthaldicarboxaldehyde, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-784-8

RTECSNumber : WZ0430000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only