missing translation for 'onlineSavingsMsg'
Learn More

Terebic Acid 98.0+%, TCI America™

Catalog No. T12085G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12085G Supplier TCI America Supplier No. T12085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-91-4
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.153
MDL Number MFCD00022485
InChI Key UZBOWOQARWWIER-UHFFFAOYSA-N
Synonym terebic acid, tetrahydro-2,2-dimethyl-5-oxo-3-furoic acid, 3-furancarboxylic acid, tetrahydro-2,2-dimethyl-5-oxo, 2,2-dimethyl-5-oxotetrahydrofuran-3-carboxylic acid, terebinic acid, 2,2-dimethyl-5-oxo-2,3,4-trihydrofuran-3-carboxylic acid, terebinsaure, 2,2-dimethyl-5-oxo-tetrahydro-furan-3-carboxylic acid, 2,2-dimethyl-5-oxotetrahydro-3-furancarboxylic acid #, 3-furancarboxylic acid,tetrahydro-2,2-dimethyl-5-oxo
PubChem CID 101540
IUPAC Name 2,2-dimethyl-5-oxooxolane-3-carboxylic acid
SMILES CC1(C(CC(=O)O1)C(=O)O)C

Specifications

Melting Point 175°C
Color White
Quantity 5 g
Formula Weight 158.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Terebic Acid