accessibility menu, dialog, popup

UserName

CAS RN 79-91-4

IUPAC Name: (3S)-2,2-dimethyl-5-oxooxolane-3-carboxylate
Synonyms: (3S)-2,2-dimethyl-5-oxooxolane-3-carboxylate
Molecular Weight (g/mol): 157.15
Molecular Formula: C7H9O4
InChi Key: UZBOWOQARWWIER-SCSAIBSYSA-M
SMILES: CC1(C)OC(=O)C[C@@H]1C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results