missing translation for 'onlineSavingsMsg'
Learn More

Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta)

Catalog No. p-4824981
Change view
Click to view available options
Quantity:
10 mm
25 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44049HF 10 mm
AA44049DH 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44049HF Supplier Thermo Scientific Chemicals Supplier No. 044049HF
Only null left

Form: Approx. 1.04g/mm; Cast rod, diameter is approximate | CAS: 7440-27-9 | Tb | 158.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Physical Form Cast Rod
Quantity 10 mm
Assay Percent Range 99.9% (Metals basis)
Appearance Silver Gray rod
Chemical Name or Material Terbium
EINECSNumber 231-137-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only