missing translation for 'onlineSavingsMsg'
Learn More

Terbium ingot, 99.9% (REO), Ta <0.4%

Catalog No. p-4782177
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0032004 2 g
AA0032009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0032004 Supplier Thermo Scientific Chemicals Supplier No. 00032004
Only null left

CAS: 7440-27-9 | Tb | 158.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Physical Form Ingot
Quantity 2 g
Assay Percent Range 99.9% (REO)
Appearance Silver Gray ingot
Chemical Name or Material Terbium, Rare Earth Oxides Content, Ta <0.2%
EINECSNumber 231-137-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only