missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog No. AA40573NC
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40573NC 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40573NC Supplier Thermo Scientific Chemicals Supplier No. 040573NC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14014-09-6

In observation of slow magnetic relaxation in triple-stranded lanthanide helicates Terbium(III) perchlorate is substituted for dysprosium(III) perchlorate hexahydrate. By reacting 3-formylchromone and hydrated terbium (III) perchlorate in isopropanol, 3-hydroxymethylchromone [icm], bis(3-chromonyl)methanol [dcm] and the novel complex Tb(imc)3(ClO4)3.5H2O are obtained.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14014-09-6
MDL Number MFCD00150362

Specifications

Color Colorless
Physical Form Liquid
Quantity 10 g
Linear Formula Tb(ClO4)3·6H2O
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 565.37 (457.28 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Density 1.5 g/mL
Chemical Name or Material Terbium(III) perchlorate
Show More Show Less
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-826-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only