accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OTbOClOOOOClOOOOClOOO

CAS RN 14014-09-6

IUPAC Name: terbium(3+) hexahydrate triperchlorate
Synonyms: terbium(3+) hexahydrate triperchlorate
Molecular Weight (g/mol): 565.35
Molecular Formula: Cl3H12O18Tb
InChi Key: NAPXFYQZNZUQDE-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Tb+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results