missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) nitrate hexahydrate, 99.99+%, (trace metal basis)

Catalog No. AC295790020
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC295790020 2 g
AC295790100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295790020 Supplier Thermo Scientific Chemicals Supplier No. 295790020

CAS: 13451-19-9 | H12N3O15Tb | 453.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13451-19-9
Molecular Formula H12N3O15Tb
Molecular Weight (g/mol) 453.03
MDL Number MFCD00799162,MFCD00149868
InChI Key UTCARTSNNKGRTD-UHFFFAOYSA-N
Synonym terbium trinitrate hexahydrate, terbium iii nitrate hexahydrate, terbium nitrate, hexahydrate, terbium iii nitrate, hexahydrate 1:3:6, terbium nitrate hexahydrate, nitric acid, terbium 3+ salt, hexahydrate, 3no3.tb.6h2o, terbium iii nitratehexahydrate, terbium 3+ trinitrate hexahydrate, terbium iii nitrate hydrate, reacton
PubChem CID 202879
IUPAC Name terbium(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Physical Form Granular Powder
Quantity 2 g
Assay Percent Range 99.99% min. (TREO base)
Linear Formula Tb(NO3)3·6H2O
Beilstein 11,9087
Merck Index 14,9157
Solubility Information Solubility in water: soluble
Formula Weight 453.02
Percent Purity ≥99.99%
Grade Trace Metal Basis
Chemical Name or Material Terbium(III) nitrate hexahydrate, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
RTECSNumber WY9625000

RUO – Research Use Only