missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

CAS: 13708-63-9 | F3Tb | 215.92 g/mol

$233.01 - $743.19

Chemical Identifiers

CAS 13708-63-9
Molecular Formula F3Tb
Molecular Weight (g/mol) 215.92
MDL Number MFCD00049572
InChI Key LKNRQYTYDPPUOX-UHFFFAOYSA-K
IUPAC Name terbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tb+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1365706
View Documents
Thermo Scientific Chemicals
01365706
5 g
Each for $233.01
Only null left
 
AA1365714
View Documents
Thermo Scientific Chemicals
01365714
25 g
Each for $743.19
Only null left
 
Description

Description

Terbium(III) fluoride is used as a source of terbium. It is also useful in the preparation of fluoride glasses. Further, it is used in the preparation of electroluminescent thin films and luminescent zinc sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13708-63-9
215.92
LKNRQYTYDPPUOX-UHFFFAOYSA-K
[F-].[F-].[F-].[Tb+3]
F3Tb
MFCD00049572
terbium(3+) trifluoride

Specifications

1172°C
Crystalline
F3Tb
UN3288
LKNRQYTYDPPUOX-UHFFFAOYSA-K
terbium(3+) trifluoride
215.92
Reacton
Hygroscopic
(REO)
13708-63-9
5 g
MFCD00049572
Slightly soluble in mineral acids. Insoluble in water.
[F-].[F-].[F-].[Tb+3]
215.92
99.9%
Odorless
Terbium(III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 237-247-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only