CAS RN 13708-63-9
IUPAC Name:
terbium(3+) trifluoride
Synonyms:
terbium(3+) trifluoride
Molecular Weight (g/mol):
215.92
Molecular Formula:
F3Tb
InChi Key:
LKNRQYTYDPPUOX-UHFFFAOYSA-K
SMILES:
[F-].[F-].[F-].[Tb+3]
1
–
1
of
1
results
1
Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
CAS: 13708-63-9 Molecular Formula: F3Tb Molecular Weight (g/mol): 215.92 MDL Number: MFCD00049572 InChI Key: LKNRQYTYDPPUOX-UHFFFAOYSA-K IUPAC Name: terbium(3+) trifluoride SMILES: [F-].[F-].[F-].[Tb+3]