missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) chloride, ultra dry, 99.99% (REO)

Catalog No. p-7050186
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3579809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3579809 Supplier Thermo Scientific Chemicals Supplier No. 03579809
Only null left

CAS: 10042-88-3

Terbium(III) chloride is used generally where Tb3+ ions can be used to check for the presence of microbes. Terbium chloride is applied to the test area, which is then illuminated with UV light. Within minutes, any live endospores present will glow green.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 10042-88-3
Molecular Formula Cl3Tb
Molecular Weight (g/mol) 265.28
MDL Number MFCD00011258
InChI Key GFISHBQNVWAVFU-UHFFFAOYSA-K
IUPAC Name trichloroterbium
SMILES Cl[Tb](Cl)Cl

Specifications

Melting Point 588°C
Physical Form Powder
Quantity 10 g
Merck Index 14,9157
Solubility Information It is soluble in water.
Formula Weight 265.29
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) chloride, Ultra dry
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-132-4
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only