accessibility menu, dialog, popup

UserName

CAS RN 10042-88-3

IUPAC Name: trichloroterbium
Synonyms: terbium(III) chloride
Molecular Weight (g/mol): 265.28
Molecular Formula: Cl3Tb
InChi Key: GFISHBQNVWAVFU-UHFFFAOYSA-K
SMILES: Cl[Tb](Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results