missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbium(III) acetate hydrate, 99.99%
CAS: 100587-92-6 | C6H9O6Tb | 336.06 g/mol
Supplier: Thermo Scientific Chemicals A1738814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Terbium(III) acetate hydrate | |
| C6H9O6Tb | |
| 25 g | |
| JQBILSNVGUAPMM-UHFFFAOYSA-K | |
| terbium(3+) triacetate | |
| 336.08 (Anhydrous) |
| 100587-92-6 | |
| MFCD00149233 | |
| Hygroscopic | |
| [Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
| 336.06 | |
| 99.99% |
Chemical Identifiers
| 100587-92-6 | |
| 336.06 | |
| JQBILSNVGUAPMM-UHFFFAOYSA-K | |
| [Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
| C6H9O6Tb | |
| MFCD00149233 | |
| terbium(3+) triacetate |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only