missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) 2,4-pentanedionate, 99.9% (REO)

Catalog No. AA4003209
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4003209 10 g
AA4003204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4003209 Supplier Thermo Scientific Chemicals Supplier No. 04003209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14284-95-8 | C15H21O6Tb | 456.25 g/mol

Terbium(III) 2,4-pentanedionate is used as an active pharmaceutical ingredient. It is also used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14284-95-8
Molecular Formula C15H21O6Tb
Molecular Weight (g/mol) 456.25
MDL Number MFCD00013503
InChI Key MAPWZFZLNRMAMO-UHFFFAOYSA-N
Synonym terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11
PubChem CID 129878174
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;terbium
SMILES [Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 456.25
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Terbium(III) 2,4-pentanedionate
EINECSNumber 238-195-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only