accessibility menu, dialog, popup

UserName

CAS RN 14284-95-8

IUPAC Name: terbium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: terbium(3+) triacetylacetonate
Molecular Weight (g/mol): 456.25
Molecular Formula: C15H21O6Tb
InChi Key: MAPWZFZLNRMAMO-UHFFFAOYSA-N
SMILES: [Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results