missing translation for 'onlineSavingsMsg'
Learn More

Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO)

Catalog No. p-4824979
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12430FF 25 x 25 mm
AA12430FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12430FF Supplier Thermo Scientific Chemicals Supplier No. 012430FF
Only null left

Form: Approx. 0.52g/25 x 25mm | CAS: 7440-27-9 | Tb | 158.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Physical Form Foil
Quantity 25 x 25 mm
Assay Percent Range 99.9% (REO)
Appearance Silver Gray foil
Chemical Name or Material Terbium
Hazard Category H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-137-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only