missing translation for 'onlineSavingsMsg'
Learn More

Terazosin Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015282
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015282 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015282 Supplier Merck Emd Millipore Supplier No. PHR17211G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Terazosin Hydrochloride belongs to the class of a1-adrenergic blockers, widely used for the treatment of benign prostatic hyperplasia (BPH) affecting the urogenital tract in men.Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 70024-40-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H25N5O4 · HCl · 2H2 O
MDL Number MFCD00941408
Synonym Terazosin hydrochloride dihydrate; 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(tetrahydro-2-furanyl)carbonyl]piperazine hydrochloride dihydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 459.92
Grade Certified Reference Material