Learn More
Greiner Bio-One Terasaki Plates
For microbatch protein crystallography. Greiner Bio-One Terasaki Plates are supplied with fitting lid.
Supplier:  Greiner Bio-One 653102
 View more versions of this product
Catalog No. 07000623
Only null left
Explore available promotions
Description
Description
  • Conical well profile
  • Flat well bottom
  • Rim of the Terasaki plates makes it possible to fill all of wells with oil at same time
  • Available with 60 and 72 wells
Specifications
Specifications
Clear
Polystyrene
11.5 L (Well)
Polystyrene
Non-sterile
11.5 mL
Lid
Closure Included
Flat
Clear
Rectangular
Microtest Plate
60
Product Suggestions
Product Suggestions