missing translation for 'onlineSavingsMsg'
Learn More

Teprenone 95.0+%, TCI America™

Catalog No. p-7137364
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27011G 1 g
T27015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27011G Supplier TCI America Supplier No. T27011G
Only null left

Chemical Identifiers

CAS 6809-52-5
Molecular Formula C23H38O
Molecular Weight (g/mol) 330.556
MDL Number MFCD00869547
InChI Key HUCXKZBETONXFO-AJDZVAQLSA-N
Synonym Geranylgeranylacetone, 6,10,14,18-Tetramethyl-5,9,13,17-nonadecatetraen-2-one
PubChem CID 53384398
ChEBI CHEBI:31649
IUPAC Name (9E,13E)-6,10,14,18-tetramethylnonadeca-5,9,13,17-tetraen-2-one
SMILES CC(=CCCC(=CCCC(=CCCC(=CCCC(=O)C)C)C)C)C

Specifications

Color Yellow
Boiling Point 160°C
Quantity 1 g
Formula Weight 330.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Teprenone [mixture of (5E,9E,13E)- and (5Z,9E,13E)- isomers]
RTECSNumber RA5391300
TSCA No
Recommended Storage Freezer