accessibility menu, dialog, popup

UserName

CAS RN 6809-52-5

IUPAC Name: (5E,9E,13E)-6,10,14,18-tetramethylnonadeca-5,9,13,17-tetraen-2-one
Synonyms: teprenone teprenonum teprenon teprenona selbex
Molecular Weight (g/mol): 330.56
Molecular Formula: C23H38O
InChi Key: HUCXKZBETONXFO-NJFMWZAGSA-N
SMILES: CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CCC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results