missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tensin 4 Recombinant Rabbit Monoclonal Antibody (23GB4225)
GREENER_CHOICE

Catalog No. MA552666
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552666 100 μL
MA552667 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552666 Supplier Invitrogen™ Supplier No. MA552666
Only null left

Rabbit Recombinant Monoclonal Antibody

Tensin 4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.

Specifications

Antigen Tensin 4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4225
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNS4
Gene Accession No. Q4V8I3, Q8BZ33, Q8IZW8
Gene Alias 9930017A07Rik; AA589547; AU016405; C terminal tensin like; CTEN; c-ten; C-terminal tensin-like protein; PP14434; RGD1310402; tensin 4; tensin-4; TNS4
Gene Symbols TNS4
Host Species Rabbit
Immunogen A synthesized peptide derived from TNS4 (200-233AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217169, 303517, 84951
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less