missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir Hydrate 98.0+%, TCI America™

Catalog No. T30061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30061G 1 g
T30065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30061G Supplier TCI America Supplier No. T30061G

Chemical Identifiers

CAS 147127-20-6
Molecular Formula C9H14N5O4P
Molecular Weight (g/mol) 287.216
MDL Number MFCD00943794
InChI Key SGOIRFVFHAKUTI-ZCFIWIBFSA-N
Synonym (R)-[[2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid
PubChem CID 464205
ChEBI CHEBI:63625
IUPAC Name [(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethylphosphonic acid
SMILES CC(CN1C=NC2=C1N=CN=C2N)OCP(=O)(O)O

Specifications

Melting Point 283°C
Color White
Quantity 1 g
Formula Weight 287.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tenofovir Hydrate
RTECSNumber SZ6563600
TSCA No
Recommended Storage Freezer