Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Foam Nitrile Coating
Knit with high-tenacity composite yarns designed to meet a range of applications. Foam nitrile coating on palm provides secure grip on wet or oily parts.
$10.22 - $11.64
Specifications
| Coating Material | Nitrile Foam |
|---|---|
| Color | Black |
| Cut Resistance Rating | ANSI A3 |
| Material | Composite Fiber |
| Product Type | Cut-resistant Gloves |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-401-240
|
Superior Glove™
S13TAFNT6 |
6 |
Pair for $11.26
Case of 240 Pair for $2,397.60
|
|
|||||
|
17-401-241
|
Superior Glove™
S13TAFNT7 |
7 |
Pair for $11.26
Case of 240 Pair for $2,397.60
|
|
|||||
|
17-401-242
|
Superior Glove™
S13TAFNT8 |
8 |
Pair for $11.64
Case of 240 Pair for $2,479.20
|
|
|||||
|
17-401-243
|
Superior Glove™
S13TAFNT9 |
9 |
Pair of 12 for $10.22
Case of 240 Pair for $2,174.40
|
|
|||||
|
17-401-244
|
Superior Glove™
S13TAFNT10 |
10 |
Pair of 12 for $11.64
Case of 240 Pair for $2,479.20
|
|
|||||
|
17-401-245
|
Superior Glove™
S13TAFNT11 |
11 |
Pair of 12 for $11.26
Case of 240 Pair for $2,397.60
|
|
|||||
Description
- Continuous filament yarn is stronger than steel on an equal-weight basis
- 1250 grams of cut protection
- Lint-free
Nitrile Palm Coat Style
- Withstands oils and solvents
- Touch screen compatible
- Black and Gray
- Ideal for handling sharp, oily glass or metal
- ANSI Cut Level A4
- High coefficient of friction
- Non-shedding
Specifications
| Nitrile Foam | |
| ANSI A3 | |
| Cut-resistant Gloves | |
| Knit |
| Black | |
| Composite Fiber | |
| Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping |