Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Foam Nitrile Coating
Knit with high-tenacity composite yarns designed to meet a range of applications. Foam nitrile coating on palm provides secure grip on wet or oily parts.
Supplier:  Superior Glove™ S13TAFNT8
 View more versions of this product
Catalog No. 17-401-242
Only null left
Explore available promotions
Description
Description
  • Continuous filament yarn is stronger than steel on an equal-weight basis
  • 1250 grams of cut protection
  • Lint-free
Nitrile Palm Coat Style
  • Withstands oils and solvents
  • Touch screen compatible
  • Black and Gray
  • Ideal for handling sharp, oily glass or metal
  • ANSI Cut Level A4
  • High coefficient of friction
  • Non-shedding
Specifications
Specifications
Nitrile Foam
ANSI A3
Cut-resistant Gloves
Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping
Black
Composite Fiber
8
Knit
Product Suggestions
Product Suggestions