Learn More
Superior Glove™ TenActiv™ 18-Gauge Composite Filament Fiber Hi-Viz Green Cut-Resistant Gloves
TenActiv™ hi-viz green composite filament fiber gloves feature a nitrile coated palm for increased grip in wet or oily conditions. Specifically designed for electrostatic dissipative (ESD) properties when working with micro-electric work.
$12.09 - $12.09
Specifications
ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Green
Cut Resistance Rating ANSI A4
Hand Specific
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
296368287
View Documents
Superior Glove™
S18TAGGFN5
5
Pair for $12.09
Pack of 12 Pair for $145.08
Only null left
296368288
View Documents
Superior Glove™
S18TAGGFN6
6
Pair for $12.09
Pack of 12 Pair for $145.08
Only null left
296368289
View Documents
Superior Glove™
S18TAGGFN7
7
Pair for $12.09
Pack of 12 Pair for $145.08
Only null left
296368290
View Documents
Superior Glove™
S18TAGGFN8
8
Pair for $12.09
Pack of 12 Pair for $145.08
Only null left
296368291
View Documents
Superior Glove™
S18TAGGFN9
9
Pair for $12.09
Pack of 12 Pair for $145.08
Only null left
296368292
View Documents
Superior Glove™
S18TAGGFN10
10
Pair for $12.09
Pack of 12 Pair for $145.08
Only null left
296368293
View Documents
Superior Glove™
S18TAGGFN11
11
Pair for $12.09
Pack of 12 Pair for $145.08
Only null left
Description
Description
  • Made with high-strength TenActiv™ yarn for impressive cut resistance
  • Foam nitrile palm coating increases grip in wet and oily conditions
  • Silicone and lint-free
  • Hi-viz green shell for better worker visibility
  • Touchscreen compatible
  • Specifically designed for electrostatic dissipative (ESD) properties when working with micro-electric work
Specifications
Specifications
ANSI 4
Green
Specific
TenActiv Yarn
Metal fabrication, construction, automotive, metal stamping, general purpose
TenActiv™
Nitrile
ANSI A4
No
Cut-resistant Gloves
ASTM F2878 LEVEL 4
18
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'