missing translation for 'onlineSavingsMsg'
Learn More

TEMED (Electrophoresis), Fisher BioReagents

Catalog No. p-28821
Change view
Click to view available options
Quantity:
26 mL
130 mL
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP15020 26 mL Amber Glass
BP150100 130 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP15020 Supplier Fisher BioReagents Supplier No. FLBP150 20
Only null left

For polymerization of acrylamide/bis-acrylamide

TEMED is a commonly used catalyst for the polymerization of acrylamide/bis-acrylamide.

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Melting Point -55°C
Color Colorless
Density 0.770g/cm³
pH 8.0 to 8.5
Boiling Point 120°C
Assay Percent Range ≥97 % (by titration)
Packaging Amber Glass
Linear Formula (CH3)2NCH2CH2N(CH3)2
Quantity 26 mL
UN Number 2372
Merck Index 15, 9277
Formula Weight 116.21
Percent Purity ≥97%
Grade DNase-, RNase- and Protease-Free
Physical Form Liquid
Chemical Name or Material TEMED
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage RT