missing translation for 'onlineSavingsMsg'
Learn More

TEMED, Approx. 99%, Molecular Biology Reagent, MP Biomedicals™

Catalog No. p-4774862
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19401925 25 mL
ICN19401980 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19401925 Supplier MP Biomedicals Inc Supplier No. 0219401925
Only null left

N,N,N',N'-Tetramethylethylenediamine is used to catalyze the formation of free radicals from ammonium persulfate or riboflavin

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Density 0.775g/mL at 20°C
Assay Percent Range Approx. 99%
Quantity 25 mL
Formula Weight 116.2
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine, FTIR
EINECSNumber 203-744-6