missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEM7 Monoclonal Antibody (197C193 (IM193))
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA141065
Description
Suggested positive control: antigen standard for PLXDC1 (transient overexpression lysate), HCT-116. Immunogen sequence: LQNNLSPKTK GTPVHLG.
Recently, using SAGE (Serial Analysis of Gene Expression) technology, St. Croix et al, have identified 46 genes, whose expression is specifically elevated in tumor-associated endothelium. Nine of these genes were prominently expressed only in tumor endothelial cells (EC), but were absent or barely detectable in normal ECs, and named as Tumor Endothelial Markers (TEMs, TEM 1-9). TEM7 (Tumor endothelial marker 7) transcripts are specifically expressed in the endothelium of colorectal cancer, primary cancers of lung, pancreas, breast, and brain. TEM7 is expressed specifically in endothelium of these cancers, whether primary or metastasis. The other six members of this family (TEM1, 3, 4, 5, 8, and 9) also show similar expression pattern in lung and brain tumors, and liver metastasis. Since most of the genes expressed differentially in tumor endothelium are also expressed during angiogenesis, these newly discovered genes might provide important resources for basic and clinical studies of human angiogenesis.Specifications
| TEM7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q8IUK5, Q91ZV7 | |
| PLXDC1 | |
| Amino acids 409-425 of human TEM7. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 197C193 (IM193) | |
| Unconjugated | |
| PLXDC1 | |
| 2410003I07Rik; ADP-ribosylation factor-like 12; AI848450; Arl12; DKFZp686F0937; FLJ36270; FLJ45632; plexin domain containing 1; plexin domain-containing protein 1; PLXDC1; TEM3; TEM7; TEM7 related; tumor endothelial marker 3; Tumor endothelial marker 7; tumor endothelial marker7 | |
| Mouse | |
| Protein G | |
| RUO | |
| 303505, 57125, 72324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |