missing translation for 'onlineSavingsMsg'
Learn More

Chiron Telomeric Alcohol Standards

Catalog No. 2847950ME
Change view
Click to view available options
Chemical Name or Material:
1H,1H,2H,2H,3H,3H-Perfluorononan-1-ol
1H,1H,2H,2H-Perfluoro-7-methyloctan-1-ol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
2847950ME 1H,1H,2H,2H-Perfluoro-7-methyloctan-1-ol
2844950ME 1H,1H,2H,2H,3H,3H-Perfluorononan-1-ol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2847950ME Supplier Chiron Supplier No. 2847.950ME
Only null left

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material 1H,1H,2H,2H-Perfluoro-7-methyloctan-1-ol
CAS 20015-46-7
Concentration 50 μg/mL
Synonym 7Me 6:1 FTOH
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL