missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chiron Telomeric Acid, FTCAs Standards
PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.
$870.11 - $870.11
Specifications
| Concentration | 50 μg/mL |
|---|---|
| Type | Reference Material |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Catalog Number | Mfr. No. | Chemical Name or Material | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Chemical Name or Material | Price | Quantity | |||||
|
85-481-050ME
|
Chiron
85481050ME |
2H,2H-Perfluorodecanoic acid |
Each for $870.11
|
|
|||||
|
15-034-850ME
|
Chiron
15034850ME |
2H,2H-Perfluorooctanoic acid |
Each for $870.11
|
|
|||||
|
15-417-1050ME
|
Chiron
154171050ME |
2H,2H,3H,3H-Perfluorodecanoic acid |
Each for $870.11
|
|
|||||
|
28-306-50ME
|
Chiron
2830650ME |
2H,2H,3H,3H-Perfluorohexanoic acid |
Each for $870.11
|
|
|||||
|
14-847-850ME
|
Chiron
14847850ME |
2H,2H,3H,3H-Perfluorooctanoic acid |
Each for $870.11
|
|
|||||
Description
- Various singles available
- Solutions in assorted solvents
- Useful for various matrices
- Custom standards of other PFAS compounds available on demand
Specifications
| 50 μg/mL | |
| Reference Material | |
| Methanol | |
| 1 mL |