missing translation for 'onlineSavingsMsg'
Learn More

Chiron Telomeric Acid, FTCAs Standards

Catalog No. 85481050ME
Change view
Click to view available options
Chemical Name or Material:
2H,2H,3H,3H-Perfluorodecanoic acid
2H,2H,3H,3H-Perfluorohexanoic acid
2H,2H,3H,3H-Perfluorooctanoic acid
2H,2H-Perfluorodecanoic acid
2H,2H-Perfluorooctanoic acid
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
85481050ME 2H,2H-Perfluorodecanoic acid
15034850ME 2H,2H-Perfluorooctanoic acid
154171050ME 2H,2H,3H,3H-Perfluorodecanoic acid
2830650ME 2H,2H,3H,3H-Perfluorohexanoic acid
14847850ME 2H,2H,3H,3H-Perfluorooctanoic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 85481050ME Supplier Chiron Supplier No. 8548.1050ME
Only null left

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material 2H,2H-Perfluorodecanoic acid
CAS 27854-31-5
Concentration 50 μg/mL
Synonym 8:2 FTCA
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL