missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chiron Telomeric Acid, FTCAs Standards
Description
- Various singles available
- Solutions in assorted solvents
- Useful for various matrices
- Custom standards of other PFAS compounds available on demand
Specifications
Specifications
| Chemical Name or Material | 2H,2H,3H,3H-Perfluorohexanoic acid |
| CAS | 356-02-5 |
| Concentration | 50 μg/mL |
| Synonym | 3:3 FTCA |
| Type | Reference Material |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |