Learn More
Thermo Scientific Chemicals Telmisartan, 98%
An angiotensin II receptor antagonist | CAS: 144701-48-4 | C33H30N4O2
$136.15 - $367.34
Chemical Identifiers
| CAS | 144701-48-4 |
|---|---|
| Molecular Formula | C33H30N4O2 |
| Synonym | BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6144103
|
Thermo Scientific Chemicals
J6144103 |
1 g |
Each for $367.34
|
|
|||||
|
AAJ61441MD
|
Thermo Scientific Chemicals
J61441MD |
250 mg |
Each for $136.15
|
|
|||||
Description
Telmisartan also acts as a partial agonist of PPARγ, activating the receptor to 25-30% of that produced by the full agonist rosiglitazone (EC50 = 4.5 ?M). Through these actions, telmisartan potently reduces blood pressure in various animal models of hypertension, diminishing cardiac hypertrophy, cardiovascular and renal risk, and glomerulosclerosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 144701-48-4 | |
| C33H30N4O2 | |
| BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid |
Specifications
| White | |
| 144701-48-4 | |
| 514.62g/mol | |
| 261°C to 263°C | |
| Powder |
| An angiotensin II receptor antagonist | |
| C33H30N4O2 | |
| BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid | |
| Signal Transduction Reagent | |
| 99% |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only