missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Telmisartan, 98%

Catalog No. AAJ6144103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6144103 1 g
AAJ61441MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6144103 Supplier Thermo Scientific Chemicals Supplier No. J6144103

An angiotensin II receptor antagonist | CAS: 144701-48-4 | C33H30N4O2

Telmisartan also acts as a partial agonist of PPARγ, activating the receptor to 25-30% of that produced by the full agonist rosiglitazone (EC50 = 4.5 ?M). Through these actions, telmisartan potently reduces blood pressure in various animal models of hypertension, diminishing cardiac hypertrophy, cardiovascular and renal risk, and glomerulosclerosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144701-48-4
Molecular Formula C33H30N4O2
Synonym BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid

Specifications

Quantity 1 g
Description White
For Use With (Application) An angiotensin II receptor antagonist
Formula Weight 514.62g/mol
Melting Point 261°C to 263°C
Product Type Signal Transduction Reagent
Form Powder
Assay 99%
Recommended Storage Ambient temperatures

RUO – Research Use Only