missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Telmisartan, 98%

An angiotensin II receptor antagonist | CAS: 144701-48-4 | C33H30N4O2

Supplier:  Thermo Scientific Chemicals J61441MD

 View more versions of this product

Catalog No. AAJ61441MD


Only null left

Description

Description

Telmisartan also acts as a partial agonist of PPARγ, activating the receptor to 25-30% of that produced by the full agonist rosiglitazone (EC50 = 4.5 ?M). Through these actions, telmisartan potently reduces blood pressure in various animal models of hypertension, diminishing cardiac hypertrophy, cardiovascular and renal risk, and glomerulosclerosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

250 mg
An angiotensin II receptor antagonist
144701-48-4
514.62g/mol
261°C to 263°C
Powder
White
99%
C33H30N4O2
BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid
Signal Transduction Reagent

Chemical Identifiers

144701-48-4
C33H30N4O2
BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only