missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tellurium AA Standard, 1000 ppm Te in 3% HCl, Ricca Chemical
$127.84 - $308.00
Chemical Identifiers
| CAS | 13494-80-9 |
|---|---|
| Molecular Formula | Te |
| Molecular Weight (g/mol) | 127.60 |
| MDL Number | MFCD00134062 |
| InChI Key | PORWMNRCUJJQNO-UHFFFAOYSA-N |
| PubChem CID | 6327182 |
| ChEBI | CHEBI:30452 |
| IUPAC Name | tellurium |
| SMILES | [Te] |
Chemical Identifiers
| 13494-80-9 | |
| 127.60 | |
| PORWMNRCUJJQNO-UHFFFAOYSA-N | |
| CHEBI:30452 | |
| [Te] |
| Te | |
| MFCD00134062 | |
| 6327182 | |
| tellurium |
Specifications
| Natural LDPE Bottle | |
| 92.0,5.28,0.09 | |
| Odorless | |
| MFCD00134062 | |
| Approximately 0°C | |
| PORWMNRCUJJQNO-UHFFFAOYSA-N | |
| tellurium | |
| 100 mL | |
| CHEBI:30452 | |
| Tellurium AA Standard |
| 13494-80-9,7647-01-0,7732-18-5,7732-18-5 | |
| 96.0,5.49,0.1 | |
| Te | |
| Colorless | |
| Standard | |
| [Te] | |
| 127.60 | |
| 6327182 | |
| 1mL = 1mg Te (1, 000ppm Te)Te in 3% HCl |
Safety and Handling
ShelfLife : 24 months