missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teicoplanin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 61036-62-2 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Synonym | Teicoplanin; Targocid; Teichomycin |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Grade | Certified Reference Material |