Learn More
MilliporeSigma™ Supelco™ Tedlar™ SCV Gas Sampling Bag with Thermogreen™ LB-2 Septa
Description
Tedlar™ is made from a high-quality 2 mil Tedlar™ film for superior inertness, impermeability, sample integrity and good recovery data.
Specifications
Specifications
| Includes | Screw Cap Valve (SCV) |
| Material | PVDF Film |
| Product Line | Tedlar |
| Quantity | 5/Pk. |
| Capacity (Metric) | 25 L |
Tedlar is a registered trademark of E. I. du Pont de Nemours and Company. Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC.