Learn More
MilliporeSigma™ Supelco™ Tedlar™ Gas Sampling Bag for EPA TCLP Method
Catalog No. 111009777
Change view
Click to view available options
Capacity (Metric):
1 L
Catalog No. Capacity (Metric)
111009777 1 L
1 options
Catalog No. 111009777 Supplier MilliporeSigma™ Supelco™ Supplier No. 30277U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tedlar™ is recommended in several US EPA methods for sampling VOCs and sulfur compounds.
Tedlar™ is made from a high-quality 2 mil Tedlar™ film for superior inertness, impermeability, sample integrity and good recovery data. Our Push Lock Valve (PLV) and seam-sealing process ensure that these bags are sturdy and virtually leak-proof, even under the most demanding sampling and shipping conditions.
Specifications
Includes Septum type Thermogreen LB-2
Material PVDF Film
Product Line Tedlar
Quantity 10/Pk.
Capacity (Metric) 1 L
Tedlar is a registered trademark of E. I. du Pont de Nemours and Company. Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC.