missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer, METTLER TOLEDO

Catalog No. 01913198
Change view
Click to view available options
pH:
1.00
12.00
12.45
13.00
3.00
6.00
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
01913198 3.00
01913197 1.00
01913188 6.00
01913190 12.00
01913194 12.45
01913195 13.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01913198 Supplier METTLER TOLEDO Supplier No. 31049545

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The technical buffer solution with provides a precise acidic calibration point for pH meters.

  • It ensures accurate and reliable measurements in highly acidic environments, making it ideal for laboratories, industrial processes, and research.
  • The 250 mL size supports multiple calibrations, aiding consistent pH accuracy and regulatory compliance.

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 3.00
Physical Form Solution
Quantity 250 mL
Solution Type Technical Buffer
Grade Laboratory