missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TEC, His Tag Recombinant Protein
Recombinant Protein
$908.00 - $12740.00
Specifications
| Accession Number | P42680 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 20mM tris with 150mM NaCl, 50% glycerol, 0.5mM EDTA, 2mM DTT, 0.05% Triton X-100 and no preservative; pH 7.5 |
| Gene ID (Entrez) | 100124696, 7006 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV3269
|
Invitrogen™
PV3269 |
10 μg |
Each for $908.00
|
|
|||||
PV4090
|
Invitrogen™
PV4090 |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain and is highly expressed in many hematopoietic cell lines. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.Specifications
| P42680 | |
| Kinase Assay | |
| 100124696, 7006 | |
| Human TEC, His Tag | |
| Purified | |
| RUO | |
| TEC | |
| TEC Kinase Family | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Dry Ice | |
| Liquid |
| See Label | |
| 20mM tris with 150mM NaCl, 50% glycerol, 0.5mM EDTA, 2mM DTT, 0.05% Triton X-100 and no preservative; pH 7.5 | |
| 76.4 kDa | |
| 7.5 | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| cytoplasmic tyrosine kinase, Dscr28C related; MGC126760; MGC126762; PSCTK4; TEC; tec protein tyrosine kinase; tyrosine-protein kinase Tec | |
| Tec | |
| Protein | |
| Human | |
| His-tag | |
| Tyrosine Kinases | |
| Baculovirus |