missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tebuconazole | |
| 103°C | |
| C16H22ClN3O | |
| 5 g | |
| tebuconazole, folicur, fenetrazole, ethyltrianol, terbuconazole, terbutrazole, etiltrianol, elite, raxil, preventol a 8 | |
| CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1 | |
| 307.82 | |
| CHEBI:83779 | |
| ≥98.0% (HPLC,T) |
| 107534-96-3 | |
| White | |
| MFCD02674797 | |
| 3077 | |
| PXMNMQRDXWABCY-UHFFFAOYNA-N | |
| 1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol | |
| 86102 | |
| 307.82 | |
| Crystalline Powder |
Chemical Identifiers
| 107534-96-3 | |
| 307.82 | |
| PXMNMQRDXWABCY-UHFFFAOYNA-N | |
| 86102 | |
| 1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol |
| C16H22ClN3O | |
| MFCD02674797 | |
| tebuconazole, folicur, fenetrazole, ethyltrianol, terbuconazole, terbutrazole, etiltrianol, elite, raxil, preventol a 8 | |
| CHEBI:83779 | |
| CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1 |
Safety and Handling
EINECSNumber : (5)-6229
RTECSNumber : XZ4803270
TSCA : No