missing translation for 'onlineSavingsMsg'
Learn More

Tebuconazole 98.0+%, TCI America™

Catalog No. T29881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29881G 1 g
T29885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29881G Supplier TCI America Supplier No. T29881G

Chemical Identifiers

CAS 107534-96-3
Molecular Formula C16H22ClN3O
Molecular Weight (g/mol) 307.82
MDL Number MFCD02674797
InChI Key PXMNMQRDXWABCY-UHFFFAOYNA-N
Synonym tebuconazole, folicur, fenetrazole, ethyltrianol, terbuconazole, terbutrazole, etiltrianol, elite, raxil, preventol a 8
PubChem CID 86102
ChEBI CHEBI:83779
IUPAC Name 1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol
SMILES CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1

Specifications

Melting Point 103°C
Color White
Quantity 1 g
UN Number 3077
Formula Weight 307.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tebuconazole
EINECSNumber (5)-6229
RTECSNumber XZ4803270
TSCA No