missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TEAD1/2/3/4 Recombinant Rabbit Monoclonal Antibody (23GB4680)

Catalog No. MA552729
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552729 20 μL
MA552728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552729 Supplier Invitrogen™ Supplier No. MA552729
Only null left

Rabbit Recombinant Monoclonal Antibody

TEAD1/2/3/4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

TEAD1, TEAD2, TEAD3, and TEAD4 are members of the TEAD (TEA domain) family of transcription factors. These factors are known to play important roles in various biological processes, including development, cell proliferation, differentiation, and organogenesis. TEAD1, TEAD2, TEAD3, and TEAD4 exhibit overlapping but also distinct expression patterns and functions. They are involved in diverse cellular processes and have been implicated in the regulation of embryonic development, tissue homeostasis, cell fate determination, and tumor progression. TEAD proteins are known to interact with various co-factors, including the transcriptional co-activators YAP (Yes-associated protein) and TAZ (transcriptional co-activator with PDZ-binding motif), which play crucial roles in the Hippo signaling pathway. The interaction between TEAD and YAP/TAZ is essential for the activation of downstream target genes involved in cell proliferation and survival.

Specifications

Antigen TEAD1/2/3/4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4680
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TEAD1
Gene Accession No. P28347
Gene Alias DTEF-1; ETF; ETFR-1; Protein GT-IIC; RTEF1; RTEF-1; TCF13; TCF-13; TEA Domain Family Member 1; TEA Domain Family Member 1 (SV40 Transcriptional Enhancer Factor); TEA Domain Family Member 2; TEA Domain Family Member 3; TEA Domain Family Member 5; TEA Domain Transcription Factor 1; TEA Domain Transcription Factor 2; TEA Domain Transcription Factor 3; TEAD; TEAD2; TEAD-2; TEAD3; TEAD-3; TEAD-4; TEAD5; TEF3; TEF4; TEF-4; TEF5; TEF-5; Transcription Factor 13; Transcriptional Enhancer Factor 1; Transcriptional Enhancer Factor 1-Related; Transcriptional Enhancer Factor 5; Transcriptional Enhancer Factor TEF-1; Transcriptional Enhancer Factor TEF-4; Transcriptional Enhancer Factor TEF-5; Transcriptional Enhancer Factor TEF-5 (DTEF-1)
Gene Symbols TEAD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TEAD1+TEAD2+TEAD3+TEAD4 (150-300AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361630, 7003
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less