missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tea Extract from Camellia sinensis, ≥80% theaflavins (theaflavin and theaflavin gallates) basis, MilliporeSigma™ Supelco™
$59.68 - $415.08
Specifications
| Physical Form | Neat |
|---|---|
| Type | Analytical Standard |
| Percent Purity | ≥80% theaflavins (theaflavin and theaflavin gallates) basis |
| Grade | Analytical Standard |
Description
Featured Industries: Food and BeveragesSpecifications
| Neat | |
| Analytical Standard | |
| ≥80% theaflavins (theaflavin and theaflavin gallates) basis | |
| Analytical Standard |