missing translation for 'onlineSavingsMsg'
Learn More

TEA, 0.2M buffer soln., pH 8.0, Thermo Scientific Chemicals

Catalog No. AAJ63793AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63793AE 100 mL
AAJ63793AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63793AE Supplier Thermo Scientific Chemicals Supplier No. J63793AE

Triethanolamine (TEA), an emulsifier, surfactant and capping agent, may be used to study its toxicity profile and mechanisms. TEA may be used in various drug delivery constructs to improve drug solubility. TEA may be used to cap CdTe nanoparticles.Triethanolamine is found are liquid laundry detergents, dishwashing liquids, general cleaners, hand cleaners, polishes, metalworking fluids, paints, shaving cream and printing inks. Triethanolamine is also used as organic additive in the grinding of cement clinker. TEA has also been used in photographic (silver halide) processing. It has been promoted as a useful alkali by amateur photographers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethanolamine (TEA), an emulsifier, surfactant and capping agent, may be used to study its toxicity profile and mechanisms. TEA may be used in various drug delivery constructs to improve drug solubility. TEA may be used to cap CdTe nanoparticles.Triethanolamine is found are liquid laundry detergents, dishwashing liquids, general cleaners, hand cleaners, polishes, metalworking fluids, paints, shaving cream and printing inks. Triethanolamine is also used as organic additive in the grinding of cement clinker. TEA has also been used in photographic (silver halide) processing. It has been promoted as a useful alkali by amateur photographers.

Solubility
Soluble in water.1 M, clear, colorless

Notes
Air Sensitive & Hygroscopic.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture, air, acids, metals, halocarbons.

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym Triethanolamine buffer solution
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

Color Colorless
Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Solubility Information Soluble aq. soln.0.1 M,clear,colorless
Chemical Name or Material TEA buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only